missing translation for 'onlineSavingsMsg'
Learn More

Celite™ S, filter aid, Honeywell Fluka™

Catalog Number p-7120304
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Fiberboard Box
Poly Bottle
This item is not returnable. View return policy
61790-53-2
O2Si
60.083
MFCD00132803
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
CHEBI:30563
dioxosilane
O=[Si]=O
This item is not returnable. View return policy
61790-53-2
O2Si
60.083
MFCD00132803
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
CHEBI:30563
dioxosilane
O=[Si]=O

Filter aid, dried, untreated

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Color Gray
Quantity 1 kg
Packaging Poly Bottle
UN Number NONH for all modes of transport
Chemical Name or Material Celite™ S
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'