missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CellEvent™ Senescence Green Flow Cytometry Assay Kit

Product Code. p-7180911
Click to view available options
Quantity:
200 Assays
50 Assays
Unit Size:
Pack of 200
Pack of 50
This item is not returnable. View return policy

Product Code. 16366622

Brand: Invitrogen™ C10841

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

The CellEvent Senescence Green Flow Cytometry Assay Kit enables flow cytometric detection of cellular senescence via β-galactosidase hydrolysis.

The kit contains an optimized buffer and a fluorescein-based reagent that contains two galactoside moieties, making it a specific target for β-galactosidase. Activation of β-galactosidase is commonly used as a biomarker for senescent cells. This hydrolase enzyme resides in lysosomes and converts β-galactosides into monosaccharides under acidic pH conditions. The enzyme-cleaved product is retained within the cell due to covalent binding of intracellular proteins and emits a fluorogenic signal that has absorption/emission maxima of 490/514 nm.

CellEvent Senescence Green Flow Cytometry Assay Kit features include:
• Fluorescent detection of β-galactosidase hydrolysis by flow cytometry
• Fixed cell assay
• Multi-color compatibility

When normal cells reach the end of their limited replicative lifespan, commonly referred to as the Hayflick limit, they enter into a senescent phase where they remain metabolically active without undergoing cell death processes. These senescent cells adopt a specific phenotypic state that includes the appearance of multi-nucleated cells, increased vacuolization, expression of pH-dependent β-galactosidase, and morphological changes where cells become enlarged and extended. Through a variety of mechanisms, they may play a role in tumor suppression, tumor progression, aging, and tissue repair.

β-galactosidase resides in lysosomes and converts β-galactosides into monosaccharides under acidic pH conditions. The activity is optimal under lysosomal pH 4, but conventional assays measure at pH 6. It has been shown that normalized β-galactosidase activity is twice as high in senescent cells as in presenescent cells regardless of the pH value used for testing.

Traditionally, the colorimetric substrate for β-galactosidase, 5-bromo-4-chloro-3-indolyl-β-D-galactopyranoside (x-gal), has long been used to detect metabolic activity in cells in vitro. Upon hydrolysis by β-galactosidase, x-gal converts to a blue precipitate, which is not suitable for flow cytometric assays. Therefore, we have developed a sensitive, fluorescent substrate for β-galactosidase that can be used in flow cytometry assays for the detection of senescent cells.

TRUSTED_SUSTAINABILITY

Specifications

Color Green
Detection Method Fluorescence
For Use With (Application) Flow Cytometry
For Use With (Equipment) Flow Cytometer
Product Type Flow Cytometry Assay Kit
Emission 488/514
Product Line CellEvent
Quantity 200 Assays
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.