Learn More
Thermo Scientific™ CellSensor™ NFAT bla HEK 293T Cell Line
Product Code. 10667363
Change view
Click to view available options
Quantity:
1 Vial
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10667363 1 Vial Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10667363 Brand Thermo Scientific™ Brand No. K1538
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
The CellSensor™ NFAT-bla HEK 293T cell line contains a beta-lactamase reporter gene under the control of the Nuclear Factor of Activated T Cells (NFAT) response element stably integrated into HEK 293T cells.
The CellSensor™ NFAT-bla HEK 293T cell line contains a beta-lactamase reporter gene under the control of the Nuclear Factor of Activated T Cells (NFAT) response element stably integrated into HEK 293T cells. Fluorescence activated cell sorting (FACS) was used to isolate clones responsive to stimulation of the NFAT pathway by PMA (Phorbol 12-Myristate 13-Acetate) and Thapsagargin (Thaps). The cell line was validated for DMSO tolerance, cell number, stimulation time, substrate loading conditions, and EC50 concentration of Thaps at fixed concentration of PMA. The CellSensor™ NFAT-bla HEK 293T cell line responds to agonist treatment as expected from the literature and can be adapted for high-throughput screening (including coupling of receptors to the NFAT pathway) for agonists or antagonists with compound libraries. Candidate drugs can also be tested for dose response against this cell line. Academic and non-profit customers, please inquire for special pricing.
TRUSTED_SUSTAINABILITY
Specifications
Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
Description CellSensor™ NFAT bla HEK 293T Cell Line, 1Vial, ∽ 2,000,000 cells/vial at 2,000,000 cells/mL Quantity of Cells, NFAT bla HEK 293T Cell Line, Fluorescent Detection Method, CellSensor™ Druggable Target, Adherent Growth Properties, Liquid Nitrogen Storage
Detection Method Fluorescent
For Use With (Application) Pharma and biopharma, drug discovery and development, target and lead identification and validation, kinase biology, kinase pathway analysis, pathway biology, cellular pathway analysis assays, cellSensor™ cellular pathway assays
Includes beta-lactamase reporter gene under the control of the nuclear factor of activated T cells (NFAT) response element stably integrated into HEK 293T cells
Product Line CellSensor
Quantity 1 Vial
Sample Type NFAT-bla HEK 293T
Target Entry PKC, PKC and calcium pathway
Type Cell Line
System Type CellSensor™
Druggable Target Signaling Pathway