Learn More
Greiner Bio-One CELLSTAR 96-well, Cell Culture-Treated, V-Shaped-Bottom Microplate
Color:Clear; Sterility:Sterile; Lid:Without Lid
Brand: Greiner Bio-One 651160
Additional Details : Weight : 0.80000kg
Description
- Compatible with automated systems
- Alphanumeric well coding
- Conforms to recommendation of American National Standards Institute (ANSI) (ANSI/SBS 1-2004)
Specifications
Standard | |
8 x 12 | |
Clear | |
96 | |
Sterile | |
Conical Bottom (V-well) | |
75 | |
100/Cs. |
Without Lid | |
Not Autoclavable | |
Polystyrene | |
25 to 340 μL | |
Cell Culture | |
4800 x g (swinging bucket rotor) | |
1 | |
-20°C to +60°C |