Learn More
Serva Electrophoresis™ Cellulase Onozuka RS from Trichoderma viride ca
Brand: Serva Electrophoresis™ 1642001
Additional Details : Weight : 0.00100kg
Specifications
2 μ/mg | |
Cellulase 2U/mg, Hemicellulase, Protease (DMC), α-amylase, Pectinase | |
1 U catalyzes the liberation of 1 μmole glucose from sodium carboxymethyl Cellulose per minute at 40°C, pH 4.5; glucose determined with alkaline copper reagent (3). | |
Cellulase »Onozuka« RS | |
Cellulase »Onozuka« RS from Trichoderma viride ca |
Trichoderma viride ca | |
4 to 5 | |
Store in a cool location | |
1 g |