missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Cellulose Nitrate (Mixed Cellulose Ester) Membrane Filters, Gridded, Grey /White 0.45μm High-Flow

Sartorius Stedim Biotech™ Cellulose Nitrate (Mixed Cellulose Ester) Membrane Filters, Gridded, Grey /White 0.45μm High-Flow are recommended for detection of yeasts and molds, particle count, and microscopy.

Brand:  Sartorius Stedim Biotech™ 130H6--47----ACN

Additional Details : Weight : 0.03000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II

 View more versions of this product

Product Code. 15922470

  • 139.00€ / Pack of 100

Estimated Shipment: 20-06-2023
to see stock.
Add to basket

Description

Description

  • Sterile, individually packaged filters
  • Save preparatory time
  • Perfect grid printing
  • Filter colors allow the best contrast to the colonies or particles
  • Outstanding microorganism recovery rate
  • High flow membranes
  • Sterilized by gamma irradiation
  • Compatible with aqueous solutions (pH 4-8), hydrocarbons and several other organic solvents
Specifications

Specifications

Gray/White Grid
0.45 μm
130
115 to 145 μm
Cellulose Nitrate (Cellulose Mixed Ester)
Sterile
100/pk.
47 mm
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions