missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CeMMEC1
GREENER_CHOICE

Product Code. 18797806
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18797806

Brand: Tocris Bioscience™ 7139/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

TAF1 inhibitor

CeMMEC1 is a TAF1 inhibitor (Kd = 1.8 μM, IC50 = 900 nM). CeMMEC1 impairs cell viability in combination with (+)-JQ1 (Cat. No. 4499) and inhibits proliferation of THP-1 and H23 lung adenocarcinoma cells.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(2,3-Dihydro-1,4-benzodioxin-6-yl)-1,2-dihydro-2-methyl-1-oxo-4-isoquinolinecarboxamide
CAS 440662-09-9
Quantity 10 mg
Target Bromodomain Inhibitors
Molecular Formula C19H16N2O4
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.