missing translation for 'onlineSavingsMsg'
Learn More

Nickel Electro™ Fixed Angle Head Rotor for Tubes

Product Code. 12883272
Click to view available options
Capacity:
12 x 1.5 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12883272

Brand: Nickel Electro™ R0001

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Nickel Electro™ Fixed Angle Head Rotor for Tubes is comprised of 12 rows for 1.5mL tubes with a max speed of 8000rpm.

  • Fixed
  • Angle head rotor
  • For tubes

Specifications

Capacity 12 x 1.5 mL
Max. Speed 8,000 rpm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.