missing translation for 'onlineSavingsMsg'
Learn More

Terracycle™ Centrifuge Tubes and Rigid Lab Plastics Zero Waste Box™
GREENER_CHOICE

Product Code. p-7249564
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17805735

Brand: Terracycle™ CS129SBE

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Recycle Centrifuge Tubes and Rigid Lab Plastics using this Zero Waste Box system. Zero Waste Box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.

Use this Zero Waste Box to recycle:

  • Any brand and type of rigid lab plastics: centrifuge tubes, plastic bottles, trays, vials and beakers.

Approximate waste units accepted:

  • Small: 200 units
  • Medium: 550 units
  • Large: 1000 units

Do not send in:

  • Tubes and rigid lab plastics that are contaminated with bodily fluids and materials considered hazardous or dangerous
  • Autoclaved material
  • Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) which presents a danger to the environment, or to people
  • Batteries, pressurized canisters, broken glass and medical waste

Important:

  • We do not collect any hazardous waste containing substances with toxic, reactive, corrosive, infectious, flammable, explosive properties or harmful to human health and the environment.
  • Please make sure that you do not mix hazardous waste with non-hazardous waste.
  • As the waste producer, it is your responsibility to classify the waste.

Specifications

Capacity (Metric) 7 kg
Capacity (English) 15 lb.
Color Beige, Green, Red, White
Dimensions (L x W x H) 25 x 25 x 46 cm
Disposable Single-use
Latex Free Yes
Size Small
Sterility Non-sterile
Autoclavable Not Autoclavable
Material Cardboard, Plastic
Product Type Recycling Box
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.