missing translation for 'onlineSavingsMsg'
Learn More

Centrophenoxine hydrochloride, 98%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J64196.06

Quantity 5g

 View more versions of this product

Product Code. 15466259

  • 40.10€ / 5g
 Restricted Item
Estimated Shipment: 01-10-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Ester of dimethylethanolamine and 4-chlorophenoxyacetic acid; activates acetylcholinesterase activity in hippocampus of aged rats
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

3685-84-5
294.17
FIVHOHCAXWQPGC-UHFFFAOYSA-N
19379
[H+].[Cl-].CN(C)CCOC(=O)COC1=CC=C(Cl)C=C1
C12H17Cl2NO3
MFCD00012533
meclofenoxate hydrochloride, centrophenoxine hydrochloride, lucidril, cerutil, meclofenoxate hcl, atsefen, helfergin, marucotol, acefen, brenal
hydrogen 2-(dimethylamino)ethyl 2-(4-chlorophenoxy)acetate chloride

Specifications

3685-84-5
MFCD00012533
14,5780
FIVHOHCAXWQPGC-UHFFFAOYSA-N
hydrogen 2-(dimethylamino)ethyl 2-(4-chlorophenoxy)acetate chloride
19379
98%
C12H17Cl2NO3
5g
meclofenoxate hydrochloride, centrophenoxine hydrochloride, lucidril, cerutil, meclofenoxate hcl, atsefen, helfergin, marucotol, acefen, brenal
[H+].[Cl-].CN(C)CCOC(=O)COC1=CC=C(Cl)C=C1
294.17
294.17
Centrophenoxine hydrochloride
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.