missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium chloride, Free-flowing, Reagent Grade, anhydrous, 99.9%, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly Bottle
Description
- ≥95% for general laboratory use.
Specifications
Melting Point | 645°C (literature) |
CAS Max % | 100.0000% |
Quantity | 1 kg |
Linear Formula | CsCl |
IUPAC Name | cesium;chloride |
Formula Weight | 168.36 |
Percent Purity | 99.9% |
Grade | Reagent |
Packaging | Poly Bottle |
Density | 3.98 g/cm3 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'