missing translation for 'onlineSavingsMsg'
Learn More

Cesium chloride, Free-flowing, Reagent Grade, anhydrous, 99.9%, Honeywell Fluka™

Catalog Number p-7111976
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
7647-17-8
ClCs
168.36
MFCD00010955
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
24293
CHEBI:63039
[Cl-].[Cs+]
This item is not returnable. View return policy
7647-17-8
ClCs
168.36
MFCD00010955
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
24293
CHEBI:63039
[Cl-].[Cs+]

Free-flowing, Reagent Grade, anhydrous, 99.9%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Melting Point 645°C (literature)
CAS Max % 100.0000%
Quantity 100 g
Linear Formula CsCl
IUPAC Name caesium(1+) chloride
Formula Weight 168.36
Percent Purity 99.9%
Grade Reagent
Packaging Poly Bottle
Density 3.98 g/cm3
Chemical Name or Material Cesium chloride, Free-flowing, Reagent Grade, anhydrous, 99.9%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'