missing translation for 'onlineSavingsMsg'
Learn More

Cesium iodide, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7048599
Click to view available options
Quantity:
25 g
100 g
7789-17-5
CsI
259.81
MFCD00010962
XQPRBTXUXXVTKB-UHFFFAOYSA-M
cesium iodide, caesium iodide, cesium monoiodide, dicesium diiodide, tricesium triiodide, cesiumiodide, cesium iodide cs2i2, cesium iodide cs3i3, unii-u1p3gvc56l
24601
cesium;iodide
[I-].[Cs+]
7789-17-5
CsI
259.81
MFCD00010962
XQPRBTXUXXVTKB-UHFFFAOYSA-M
cesium iodide, caesium iodide, cesium monoiodide, dicesium diiodide, tricesium triiodide, cesiumiodide, cesium iodide cs2i2, cesium iodide cs3i3, unii-u1p3gvc56l
24601
cesium;iodide
[I-].[Cs+]

Cesium iodide is used as an input phosphor in x-ray image intensifier tube of fluoroscopy equipment. It is also used as an equipment for electromagnetic calorimetry in experimental particle physics. It is often employed as a beamsplitter in Fourier Transform Infrared (FT-IR) spectrometers. Cesium iodide is an inorganic scintillator which is mainly used in experimental nuclear physics, high energy physics, nuclear medicine, nuclear tomography, environmental studies and many other fields of use. It is utilized in calorimeters and detectors and also employed as a phosphor in fluoroscopy equipments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1280°C
Melting Point 626°C
Color White
Physical Form Crystalline
Quantity 25 g
Merck Index 14,2014
Solubility Information Soluble in water. Soluble in water and ethanol. Slightly soluble in methanol. Insoluble in acetone.
Formula Weight 259.81
Percent Purity 99.999%
Odor Odorless
Refractive Index 1.7876
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Cesium iodide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'