Learn More
OHAUS™ Chain Clamp
Shop All Ohaus Products
Click to view available options
Size:
Small
Large
Material:
Nickel-plated
Stainless Steel
Length (Metric) Arm:
127 mm
305 mm
Unit Size:
Each
Descrizione
- Corrosion-resistant stainless steel clamps provide excellent chemical resistance, durability, and can be autoclaved. Nickel-plated zinc clamps offer high-tensile strength at an economical price.
- Prevent accidents in the lab! Versatile multi-purpose LabJaws clamps securely hold glassware and irregularly-shaped objects. Select from a variety of grip sizes, prong styles and adjustment types.
- Choose from a variety of specialized LabJaws clamps with adjustable features to provide an exact fit for lab ware such as thermometers, thermocouples, columns, funnels, and round bottom flasks.
Specifica
Specifica
| Type | Chain Clamp |
| Material | Nickel-plated |
| Description | Clamp, Specialty, Chain, CLS-CHAINZM |
| Diameter (Metric) | 13 mm |
| Length (English) | 7.40 in. |
| Length (English) Arm | 5.000 in. |
| Length (Metric) | 188.00 mm |
| Length (Metric) Arm | 127 mm |
| Size | Small |
| Size (Metric) Grip | 35 to 170 mm |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto