Learn More
Invitrogen™ Chargeswitch™ Pro Filter Plasmid Maxi Kit
The Chargeswitch™ -Pro Filter Plasmid Maxi Kit includes 10 plasmid maxipreps and provides high yield and high purity plasmid with the fastest and the simplest protocol.
Brand: Invitrogen™ CS31106
Additional Details : Weight : 1.14000kg
Description
The Chargeswitch™ -Pro Filter Plasmid Maxi Kit includes 10 plasmid maxipreps and provides high yield and high purity plasmid with the fastest and the simplest protocol.
The ChargeSwitch™-Pro Plasmid Filter Mini, Midi, Maxi Kits generates high-quality plasmid DNA without the use of harsh or potentially interfering reagents, for better success in downstream applications. The system is based upon a novel dual/nested column design which facilitate the fastest and simplest protocol for high quality plasmid.
The system is composed of two columns. The inner flat bottomed column, also referred to as “lysate clarification” column, contains filter materials suitable for rapid clarification of bacterial lysate. The outer luer bottomed column, also referred to as “binding” column, contains ChargeSwitch™ (CST) derivatized membrane that immediately binds plasmid DNA from the clarified lysate. This nested-column design enables simultaneous clarification and DNA binding in a single short centrifugation (or vacuum) step. The combined advantages of novel, optimized plastics and a unique chemistry are an extreme competitive advantage of speed and convenience.
To accomplish this, the ChargeSwitch™-Pro Filter Kit provides:
• 100% ethanol-free, guanidinium-free, organics-free, aqueous protocol for a safer, more streamlined method
• Cleaner purified product for improved performance in downstream applications
• A novel Dual/nested column design for simultaneous lysate clearing and binding step which offers the fastest and the simplest protocol
• Endotoxin units less than 10 EU/ug DNA for
• Vacuum Elution can be done with the EveryPrep™ Universal Vacuum Manifold (Cat. No. K211101)
How it works
The ChargeSwitch™ technology replaces the use of ethanol, chaotropic salts, or organic solvents for nucleic acid purification with a simple ion-exchange mechanism. At low pH values (less than 6.5) the ChargeSwitch™ surface ligand is positively charged and binds plasmid DNA. Raising the pH (greater than 8.5) neutralizes the charge and elutes the bound nucleic acid. The ChargeSwitch™ protocol retains the ease and familiarity of the common silica spin column protocol with a binding, washing, and elution step, making adapting to the ChargeSwitch™ method effortless.
Specifications
Spin Column | |
Spin Column | |
Maxi | |
1 to 2 mL | |
Next-Generation Sequencing, Transfection, Cloning, Sequencing, Transformation, Nucleic Acid Labeling, PCR, In Vitro Transcription | |
10 Preps | |
ChargeSwitch™ | |
10 Preps | |
Room Temperature | |
ChargeSwitch™ | |
800 μg |
The ChargeSwitch™-Pro Plasmid Kit contains Columns with Resuspension Buffer, RNase A, Lysis Buffer, Precipitation Buffer, Wash Buffers 1 and 2, and Elution Buffer. Store at room temperature. Guaranteed stable for 6 months when properly stored. | |
ChargeSwitch Chemistry | |
Bacterial Culture | |
Plasmid DNA | |
Not High-throughput Compatible (Manual) | |
> 200 μg (Large-Scale) Plasmid DNA | |
Plasmid Maxi Kit | |
Transfection Grade | |
Plasmid DNA | |
30 min. | |
100 ml |
For Research Use Only. Not for use in diagnostic procedures.