Learn More
Lovibond™ Checkit™ Comparitor Test Kits
Perform water analysis quickly and easily. Lovibond Checkit™ Comparitor Test Kits are highly accurate and incorporate a user-friendly colorimetric scale. Each kit indicates a combination of pH and another measure.
Brand: Lovibond™ 147330
Additional Details : Weight : 0.25000kg
Description
Perform water analysis quickly and easily. Lovibond Checkit™ Comparitor Test Kits are highly accurate and incorporate a user-friendly colorimetric scale. Each kit indicates a combination of pH and another measure.
- Easy operation and exact reagent dosing
- Tablet reagents with a minimm guaranteed shelf life of 5/10 years
- Rotate continuous gradient color wheel to obtain a color match with the reacted sample
- Printed continuous gradient color discs provide accurate measurements without "exrapolating" between discrete color blocks
Specifications
Chlorine dioxide | |
0.01 to 0.2 mg/L | |
Colorimetric | |
Water test kit |