missing translation for 'onlineSavingsMsg'
Learn More

Aqualytic™ Checkit Nitrate Test Kit

Aqualytic Checkit Nitrate Test Kit, Range: 0 to 1mg/L, Accuracy: +/-5%, Quantity: 1Pack

Brand:  Aqualytic™ 147810

Additional Details : Weight : 0.26200kg Transport : UN number : 3077 Chem class : 9 Pack group : III


Product Code. 11759129

  • 127.00€ / Each
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Aqualytic Checkit Nitrate Test Kit, Range: 0 to 1mg/L, Accuracy: +/-5%, Quantity: 1Pack

Specifications

Specifications

±5%
1 Pack
0 to 1mg/L
Checkit Nitrate Test Kit
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • TESTPAK CHECKIT NITRATE 0,1-1MG

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ammonium chloride, zinc powder, sulphanilic acid, sodium hydrogen sulphate

SDS
Documents

Documents

Certificates
Promotions

Promotions