Learn More
Masterflex™ Chem-Durance Bio L/S™ Two-Stop Pump Tubing
Click to view available options
Tubing Size:
13
16
25
Unit Size:
Pack of 8
Description
- Meets USP Class VI requirements
- Flexible outer jacket acts as an insulating layer to absorb and dissipate heat from the rollers - extending the life of the tubing
- Plasticizer-free, smooth bore liner is hydrophobic and resists the absorption/adsorption of aqueous fluids
Specifications
Specifications
| Diameter (Metric) Inner | 0.8 mm |
| Material | Chem-Durance Bio |
| Min. Operating Temperature (Metric) | -60°C |
| Packaging Quantity | 1 Each |
| Tubing Size | 13 |
| Flow Rate | 0.06 to 36 mL/min. |
| Hose Barb | 1/16 in. |
| Height (Metric) | 8.8 m |
| Maximum Pressure | 40 psi |
| Vacuum Level | 660 mm |
| Show More |