Learn More
Masterflex™ Chem-Durance Bio L/S™ Two-Stop Pump Tubing
Product Code. 15323174
Click to view available options
Tubing Size:
13
16
25
Unit Size:
Pack of 8
Product Code. 15323174
Brand: Masterflex™ 9611725
Only null left
Add to basket
Edge
Add to basket
Excellent chemical resistance and long pumping life
  • Meets USP Class VI requirements
  • Flexible outer jacket acts as an insulating layer to absorb and dissipate heat from the rollers - extending the life of the tubing
  • Plasticizer-free, smooth bore liner is hydrophobic and resists the absorption/adsorption of aqueous fluids
Specifications
Diameter (Metric) Inner 4.8 mm
Material Chem-Durance Bio
Min. Operating Temperature (Metric) -60°C
Packaging Quantity 1 Each
Tubing Size 25
Flow Rate 1.7 to 1000 mL/min.
Hose Barb 3/16 in.
Height (Metric) 8.8 m
Maximum Pressure 35 psi
Vacuum Level 660 mm
Max. Operating Temperature (Metric) 74°C
Product Type 2-stop Tubing