Learn More
CHEMetrics COD Vials Kits
Contains premeasured reagents for accurate, economical determination of COD
Brand: CHEMetrics K7370S
Additional Details : Weight : 0.55800kg Transport : UN number : 1830 Chem class : 8 Pack group : II
Description
- Contains everything needed (except distilled water) to perform tests
- Includes vials and instruction book
- Comes in cardboard box
- 2-hour digestion time
- Dichromate reactor digestion
A fresh reagent ampule blank must be prepared for each series of tests; therefore the number of samples that can be tested with each kit will vary.
Certifications
Not USEPA Approved
Specifications
Chemical Oxygen Demand (COD) | |
Not USEPA approved | |
Digestor block as well as a photometer or spectrophotometer | |
24 | |
Dichromate Reactor Digestion | |
COD Vials Kit |
Not USEPA approved | |
Premeasured reagents for accurate, economical determination of COD, determination of the oxidizable organic matter content of wastewater | |
25 vials and instruction book | |
0 to 15,000 ppm (HR+) | |
2 hr. digestion |
Safety and Handling
- chemical oxygen demand (COD) vials
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Germ cell mutagenicity Category 1B
- Oxidising liquid Category 3
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1A
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H272-May intensify fire; oxidiser.
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H360-May damage fertility or the unborn child.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sulphuric acid, silver sulphate, mercuric sulphate, potassium dichromate