Learn More
VACUUBRAND™ Chemistry-HYBRID Pump RC 6
GREENER_CHOICE
Product Code. 17749868
Click to view available options
Plug Type:
CEE-Plug
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 17749868
Brand: VACUUBRAND™ 20798560
Only null left
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
The RC 6 chemistry-HYBRID pump is a combination of a rotary vane pump and a chemistry diaphragm pump for optimized corrosion resistance. The diaphragm pump maintains the oil reservoir under vacuum in order to keep the partial pressures of solvent vapors at levels below their condensation points and to reduce the concentration of corrosive gases.
  • Reduced internal corrosion, even when working with corrosive vapors
  • Oil changes typically reduced 90% or more compared with rotary vane pumps alone
  • Most economical solution: In practical operation a cold trap is often no longer necessary
  • Ease of maintenance due to telescopic design
TRUSTED_SUSTAINABILITY
Specifications
Max. Vacuum 2 x 10-3 mbar
Plug Type CEE-Plug
Flow Rate 5.9 m3/hr
Pump Type Vacuum System
Enclosure Rating IP40
Maximum Pressure 1.1 bar
Voltage 230 V