Learn More
Heathrow Scientific™ Chest Freezer Rack, Stainless Steel
Shop All Thermo Scientific Incubateurs à CO2 Products
Click to view available options
Array:
13 x 1
5 x 1
8 x 1
9 x 1
Unit Size:
Each
Description
- Chest Freezer Rack
- Rack has handle for easy transfer
- Robust mounting brackets assure handles stay securely in place for easy use
- Locking Rod Keeps tube storage boxes safely in place
Specifications
Specifications
| Description | Chest Freezer Rack |
| Array | 13 x 1 |
| Color | Silver |
| Holds | Tube Boxes |
| Material | Stainless Steel |
| Sterility | Non-sterile |
| For Use With (Application) | Tube Boxes |
| Model | HS2862AE |
| No. of Racks | 13 |
| No. per Pack | 1 Ea. |
| Show More |