Learn More
Heathrow Scientific™ Chest Freezer Rack, Stainless Steel
GREENER_CHOICE
Product Code. 16330766 Shop All Thermo Scientific Incubateurs à CO2 Products
Click to view available options
Array:
13 x 1
5 x 1
8 x 1
9 x 1
Unit Size:
Each
Product Code. 16330766
Brand: Heathrow Scientific™ HS2862DB
Only null left
Add to basket
Add to basket
The Chest Freezer Rack safely stores samples in your freezer. Constructed of corrosion-resistant stainless steel racks that are compatible with major brands of chest freezers.
  • Chest Freezer Rack
  • Rack has handle for easy transfer
  • Robust mounting brackets assure handles stay securely in place for easy use
  • Locking Rod Keeps tube storage boxes safely in place
Specifications
Product Type Freezer Rack
Description Chest Freezer Rack
Array 9 x 1
Autoclavable Not Autoclavable
Color Silver
Holds Tube Boxes
Material Stainless Steel
Positions 1
Shape Rectangular
Shelves 9
Stackable No
Sterility Non-sterile
For Use With (Application) Tube Boxes
Model HS2862DB
No. of Racks 9/Pk.
No. per Pack 1 Ea.
Packaging Box
Length (Metric) 14.1 cm
Length (English) 5.5 in.
Width (Metric) 14.1 cm
Width (English) 5.5 in.
Height (Metric) 73 cm
Height (English) 29 in.