Learn More
Emma Safety Footwear Chicago Safety Shoes
Product Code. 15998391
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
Product Code. 15998391
Brand: Emma Safety Footwear MM40264742
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Light and comfortable with knitted textile upper and Nano Carbon Composite protective nose. The TPU GripForce™ Easy Twist outsole offers maximum grip, while The integrated lacing prevents pressure points from hooks and eyes and the Hydro-Tec™ lining provides optimal breathability.
  • S1P-Safety shoe with sole plate for dry work
  • Knitted Textile construction
  • Polyurethane coated
  • Hydro-Tec™ Antibacterial
  • Metal-free
  • Chrome-free lining
  • ESD Certified
Specifications
Certifications/Compliance EN ISO 20345:2011
Closure Type Laces
Color Gray, Black
Design Chicago
Liner Material Hydro-Tec™ Antibacterial
Material Textile
Size 42
Toe Material Composite