Learn More
Chicken anti-Rat IgG (H+L) Secondary Antibody, HRP, Novus Biologicals™
Chicken Polyclonal Antibody
Brand:  Novus Biologicals NBP1-75360
Product Code. 18210883
  • 324.00€ / 1mg
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG (H+L)
Polyclonal
HRP
Chicken
Affinity purified
RUO
Secondary
Rehydrate with 1.1 ml of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily.
Store lyophilized antibody at 4°C. Mix reconstituted liquid with an equal volume of glycerol, aliquot contents and freeze at -20°C or below for long term storage.
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
LYOPH
Western Blot 1:200-1:5000, ELISA 1:200-1:5000, Immunohistochemistry 1:500-1:5000, Immunohistochemistry-Paraffin
Purified Rat IgG (H&L)
1 mg
Adaptive Immunity, Immunology
Based on IEP, this antibody reacts with heavy gamma chains on rat IgG and light chains on all rat immunoglobulins
Rat