missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorambucil Isopropyl Ester, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Chlorambucil Isopropyl Ester
Specifications
Specifications
| Chemical Name or Material | Chlorambucil Isopropyl Ester |
| CAS | 77063-20-8 |
| Analyte or Component Names | Chlorambucil Isopropyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H25 Cl2 N O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Chlorambucil Isopropyl Ester, 4-[bis(2-chloroethyl)amino]-Benzenebutanoic acid 1-methylethyl ester, Chlorambucil isopropyl ester |
| InChI Formula | InChI=1S/C17H25Cl2NO2/c1-14(2)22-17(21)5-3-4-15-6-8-16(9-7-15)20(12-10-18)13-11-19/h6-9,14H,3-5,10-13H2,1-2H3 |
| SMILES | CC(C)OC(=O)CCCc1ccc(cc1)N(CCCl)CCCl |
| IUPAC Name | propan-2-yl 4-[4-[bis(2-chloroethyl)amino]phenyl]butanoate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction