missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Chlorazol Black E
124.00€
Chemical Identifiers
| CAS | 1937-37-7 |
|---|---|
| Molecular Formula | C34H25N9Na2O7S2 |
| Molecular Weight (g/mol) | 781.73 |
| MDL Number | MFCD00066363 |
| InChI Key | OLVNAWLXZDRGPL-UHFFFAOYSA-L |
| Synonym | Azo Black; C.I. 30235 |
| PubChem CID | 44135447 |
| IUPAC Name | disodium;4-amino-3-[[4-[4-[(2,4-diaminophenyl)diazenyl]phenyl]phenyl]diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate |
| SMILES | C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)C5=CC=C(C=C5)N=NC6=C(C=C(C=C6)N)N)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+] |
Description
Chemical Identifiers
| 1937-37-7 | |
| 781.73 | |
| OLVNAWLXZDRGPL-UHFFFAOYSA-L | |
| 44135447 | |
| C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)C5=CC=C(C=C5)N=NC6=C(C=C(C=C6)N)N)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+] |
| C34H25N9Na2O7S2 | |
| MFCD00066363 | |
| Azo Black; C.I. 30235 | |
| disodium;4-amino-3-[[4-[4-[(2,4-diaminophenyl)diazenyl]phenyl]phenyl]diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate |
Specifications
| 1937-37-7 | |
| MFCD00066363 | |
| OLVNAWLXZDRGPL-UHFFFAOYSA-L | |
| disodium;4-amino-3-[[4-[4-[(2,4-diaminophenyl)diazenyl]phenyl]phenyl]diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate | |
| 44135447 | |
| Chlorazol Black E |
| C34H25N9Na2O7S2 | |
| Azo Black; C.I. 30235 | |
| C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)C5=CC=C(C=C5)N=NC6=C(C=C(C=C6)N)N)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+] | |
| 781.73 | |
| 781.73 |
Safety and Handling
GHS H Statement
H350-H361
May cause cancer.
Suspected of damaging fertility or the unborn child.
P201-P202-P281-P308+P313-P501c
H350-H361d
missing translation for 'einecsNumber' : 217-710-3
missing translation for 'rtecsNumber' : QJ6160000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title