missing translation for 'onlineSavingsMsg'
Learn More

Chemetrics™ Chloride Titrets™ Titration Test Kit

Product Code. 11727413
Click to view available options
Range:
250 to 2500ppm
50 to 500ppm
Unit Size:
Pack of 30
This item is not returnable. View return policy

Product Code. 11727413

Brand: Chemetrics™ K2050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemetrics™ Chloride Titrets™ Titration Test Kit contains everything needed to perform 30 mercuric nitrate method tests for chloride.

  • Complete kit for analysis of chloride content in water
  • Applications and industries include drinking water, surface waters, seawater, domestic and industrial wastewater
  • Employs mercuric nitrate method
  • Dependable, sealed single-use titrants
  • Fast, easy procedure, with no need to count drops
  • No burettes or other equipment necessary
  • Perform a titration anywhere

Specifications

Includes Kit comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampules with valve assemblies, Activator Solution, titrettor, 25mL sample cup, and instructions
No. of Tests 30
Range 50 to 500ppm
Test Method Mercuric Nitrate
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • TITRETS CHLORIDE 50-500 MG/L
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Skin sensitiser Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.,Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.,Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P273-Avoid release to the environment.,Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.,Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.,IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302-if on skin: getmedicalhelp.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.,IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.,IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P363-Wash contaminated clothing before reuse.
  • P390-Absorb spillage to prevent material damage.
  • P405-Store locked up.,Store locked up.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.,Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contains : mercuric nitrate, nitirc acid, diethylene glycol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.