Learn More
Chemetrics™ Chloride Titrets™ Titration Test Kit
Product Code. 11737413
Click to view available options
Range:
250 to 2500ppm
50 to 500ppm
Unit Size:
Pack of 30
Product Code. 11737413
Brand: Chemetrics™ K2051
Chemetrics™ Chloride Titrets™ Titration Test Kit contains everything needed to perform 30 mercuric nitrate method tests for chloride.
  • Complete kit for analysis of chloride content in water
  • Applications and industries include drinking water, surface waters, seawater, domestic and industrial wastewater
  • Employs mercuric nitrate method
  • Dependable, sealed single-use titrants
  • Fast, easy procedure, with no need to count drops
  • No burettes or other equipment necessary
  • Perform a titration anywhere
Specifications
Includes Kit comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampules with valve assemblies, Activator Solution, titrettor, 25mL sample cup, and instructions
No. of Tests 30
Range 250 to 2500ppm
Test Method Mercuric Nitrate
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • TITRETS CHLORIDE 250-2500 MG/L
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P363-Wash contaminated clothing before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : mercuric nitrate, nitric acid, ethyleneglycol, gentisic acid