missing translation for 'onlineSavingsMsg'
Learn More

Chlorodimethylvinylsilane, 97%

Product Code. p-7046752
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11312937

Brand: Thermo Scientific Alfa Aesar L16198.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1719-58-0 | C4H8ClSi | 119.64 g/mol

Chlorodimethylvinylsilane is used to prepare silicon-containing polymers, silaheterocycles and new chelating ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorodimethylvinylsilane is used to prepare silicon-containing polymers, silaheterocycles and new chelating ligands.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive to moisture. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1719-58-0
Molecular Formula C4H8ClSi
Molecular Weight (g/mol) 119.64
MDL Number MFCD00018090
InChI Key RABBDIBWJVOAPB-ONEGZZNKSA-N
Synonym chlorodimethylvinylsilane, dimethylvinylchlorosilane, vinyldimethylchlorosilane, chloro dimethyl vinylsilane, silane, chloroethenyldimethyl, chlorodimethyl vinyl silane, unii-xja6n5221t, chloroethenyldimethyl-silane, chloro ethenyl dimethylsilane, chloro-dimethyl-ethenyl-silane
PubChem CID 519368
SMILES C[Si](C)\C=C\Cl

Specifications

Boiling Point 82°C to 83°C
Quantity 25 g
UN Number UN2985
Beilstein 1737688
Flash Point −12°C (10°F)
Solubility Information Reacts with water.
IUPAC Name chloro-ethenyl-dimethylsilane
Formula Weight 120.66
Percent Purity 97%
Refractive Index 1.416
Sensitivity Moisture sensitive
Density 0.883
Chemical Name or Material Chlorodimethylvinylsilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Chlorodimethylvinylsilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Chlorodimethylvinylsilane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.