missing translation for 'onlineSavingsMsg'
Learn More

Chloromethanesulfonyl chloride, 95%

Product Code. 11384228
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
This item is not returnable. View return policy

Product Code. 11384228

Brand: Thermo Scientific Alfa Aesar 041486.04

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 3518-65-8 | CH2Cl2O2S | 148.99 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff. It is synthesis of chloromethanesulfonates and chloromethanesulfonamides. It is also used for conversion of alcohols into the inverted acetates, azides and nitriles, and rearrangement of carbon-carbon and carbon-oxygen bonds under mild conditions, and in electrophilic and radical reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3518-65-8
Molecular Formula CH2Cl2O2S
Molecular Weight (g/mol) 148.99
MDL Number MFCD00014727
InChI Key KQDDQXNVESLJNO-UHFFFAOYSA-N
Synonym methanesulfonyl chloride, chloro, chloromethylsulfonyl chloride, chloromethanesulphonyl chloride, chlormethansulfochlorid, chlormethansulfonylchlorid, chloromesyl chloride, chloromethyl sulfonyl chloride, chlormethansulfochlorid czech, chlormethansulfonylchlorid czech, chlorid kyseliny chlormethansulfonove
PubChem CID 77054
IUPAC Name chloromethanesulfonyl chloride
SMILES ClCS(Cl)(=O)=O

Specifications

Color Colorless to Yellow
Density 1.67 g/mL
Boiling Point 78°C to 81°C (20 mmHg)
Assay Percent Range 98%
Odor Acidic
Linear Formula ClCH2SO2Cl
Quantity 2 g
UN Number UN3265
Beilstein 1811761
Sensitivity Moisture sensitive
Solubility Information It is soluble in dichloromethane,pyridine,and most organic solvents. It is insoluble in water.
Formula Weight 149
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Chloromethanesulfonyl chloride
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.