missing translation for 'onlineSavingsMsg'
Learn More

Chloromethyl phenyl sulfide, 97%, Thermo Scientific Chemicals

Catalog Number p-4228525
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
7205-91-6
C7H7ClS
158.64
MFCD00000921
LLSMWLJPWFSMCP-UHFFFAOYSA-N
chloromethyl phenyl sulfide, chloromethylthiobenzene, alpha-chlorothioanisole, phenyl chloromethyl sulfide, chloromethyl phenyl sulphide, phenylthio methyl chloride, chloromethyl thio benzene, sulfide, chloromethyl phenyl, chloromethyl phenyl sulfane, benzene, chloromethyl thio
81623
chloromethylsulfanylbenzene
ClCSC1=CC=CC=C1
This item is not returnable. View return policy
7205-91-6
C7H7ClS
158.64
MFCD00000921
LLSMWLJPWFSMCP-UHFFFAOYSA-N
chloromethyl phenyl sulfide, chloromethylthiobenzene, alpha-chlorothioanisole, phenyl chloromethyl sulfide, chloromethyl phenyl sulphide, phenylthio methyl chloride, chloromethyl thio benzene, sulfide, chloromethyl phenyl, chloromethyl phenyl sulfane, benzene, chloromethyl thio
81623
chloromethylsulfanylbenzene
ClCSC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.2300g/mL
Boiling Point 66°C (0.3 hPa)
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H5SCH2Cl
Refractive Index 1.595
Quantity 5 g
Beilstein 06,III,1002
Fieser 09,107; 06,571; 07,357; 08,472; 09,451; 10,384; 11,503; 12,468; 13,34; 14,177; 16,98; 17,327
Specific Gravity 1.23
Formula Weight 158.65
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Chloromethyl phenyl sulfide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'