Learn More
Berkshire™ Choice™ Cleanroom Face Masks
Shop All Berkshire Products
missing translation for 'orderingAttributeHoverText'
Packaging Type:
Bulk Packaged
Description
- 3-Layer cleanroom masks
- Knitted nylon earloops
- Filter medium: meltblown polypropylene provides particle and bacterial filtration
- Fully enclosed nosepiece is adjustable
- Available in bulk or individually wrapped packaging
Specifications
Specifications
| Product Type | Cleanroom Face Mask |
| Color | White |
| Fastening Type | Earloops |
| Material | Polypropylene |
| Sterility | Non-sterile |
| Style | Pleated |
| Certifications/Compliance | ASTM F2299 |
| Certifications/Compliance (Europe) | EN 14683 |
| Cleanroom Class | Class 10 (ISO 4) |
| Construction | 3-ply |
| Show More |