missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ Chopping Board with 6 HACCP Color Pellets

Chopping Board with 6 HACCP Color Pellets is ideal for intensive use. Excellent resistance to abrasion and high temperatures.

Brand:  Gilac™ G651060

Additional Details : Weight : 4.80000kg

 View more versions of this product

Product Code. 17988411

  • 52.49€ / Each
Estimated Shipment: 03-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Chopping board in HDPE, 2 cm thick, Ideal for intensive use.
  • Excellent resistance to abrasion and high temperatures.
  • Multi-purpose and easy to clean.
  • Does not retain odours from the food cut on it.
  • Its smooth surface limits knife marks and allows the blades to slide well.
  • Each board comes with a bag of 6 HACCP pellets to differentiate uses by food groups and avoid cross-contamination.
  • The pellet in the desired color is to be slotted into the board.
  • Note: once clipped, it is not possible to remove it easily, its use is designed to be dedicated to each board.
Specifications

Specifications

Cutting Board
Polyethylene
White
600 x 400 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions