missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Chorionic Gonadotropin Human/hCG Ab-5 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Chorionic Gonadotropin, Human/hCG Ab-5, Mouse Monoclonal Antibody.

134.64€ - 418.20€

Specifications

Antigen Chorionic Gonadotropin, Human/hCG Ab-5
Clone CG04 + CG05
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 6
Product Code Brand Form Quantity Price Quantity & Availability  
Product Code Brand Form Quantity Price Quantity & Availability  
12668796
View Documents Promotion Details
Epredia™
MS-1065-P1
Concentrated 500 μL
261.00€
500µL
View alternative products
Save Up to 
»
Estimated Shipment: 16-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
12608806
View Documents Promotion Details
Epredia™
MS-1065-R7
Prediluted 7 mL
204.00€
7mL
View alternative products
Save Up to 
»
Estimated Shipment: 16-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
12688796
View Documents Promotion Details
Epredia™
MS-1065-PABX
Concentrated 200 μL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
12658796
View Documents Promotion Details
Epredia™
MS-1065-P0
Concentrated 100 μL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
12648796
View Documents Promotion Details
Epredia™
MS-1065-P
Concentrated 1 mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
12678796
View Documents Promotion Details
Epredia™
MS-1065-P1ABX
Concentrated 100 μL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description

Description

Human Chorionic Gonadotropin (HCG) is a glycoprotein that is secreted in large quantities by normal trophoblasts. It is present only in trace amounts in non-pregnant urine and sera but rises sharply during pregnancy. HCG is composed of two non-identical, non-covalently linked polypeptide chains designated as the α- and Β-subunits. The α-subunit of HCG is nearly identical to that of thyroid stimulating hormone (TSH), follicle stimulating hormone (FSH), and luteinizing hormone (LH). A germ cell tumor which is positive for cytokeratin, placental alkaline phosphatase (PLAP), and HCG but negative for EMA and AFP is probably a choriocarcinoma.

Host Species: Mouse

Clone: CG04 + CG05

Isotype: IgG2a/κ + IgG1/κ

Species Reactivity: Human. Others not known

Immunogen: Purified β-subunit of hCG

Molecular Weight: 22 kDa (β-hCG)

Positive Control: Placenta or chorioca

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not optimum)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Chorionic Gonadotropin, Human/hCG Ab-5
Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human
IgG1 κ
CG04 + CG05
Monoclonal
Mouse
Steroids Hormones and Growth Factors
Purified Beta-subunit of HCG
Primary
Videos
SDS
Documents

Documents

Product Certifications