Learn More
Minigrip™ Resealable Write-on Bags
Product Code. 11961526
Encompass
Click to view available options
Width (Metric):
40 mm
75 mm
100 mm
140 mm
190 mm
Height (Metric):
65 mm
85 mm
140 mm
190 mm
Unit Size:
Pack of 100
Pack of 1000
Product Code. 11961526
Brand: Minigrip™ BAJ340003N
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Label bag contents with ease. Minigrip™ Resealable Write-on Bags provide three white bars specifically designed for writing on with inkpen or marker.
  • Polyethylene
  • Resealable
  • Autoclavable
  • Three write-on bars for labelling
Specifications
Description Resealable Bag
Color Clear
For Use With (Application) Ideal for storing stationery and sending samples
Material Polyethylene
Quantity 100 Pack
Width (Metric) 40 mm
Height (Metric) 65 mm