Learn More
Macherey-Nagel™ Chromabond™ Glass Fiber Frit
Product Code. 10347931 Shop All Macherey-Nagel Products
Click to view available options
Volume (Metric):
3 mL
6 mL
Unit Size:
Pack of 250
Product Code. 10347931
Brand: Macherey-Nagel™ 730192
Only null left
Add to basket
Add to basket
Ideal for high temperature SPE applications. Macherey-Nagel™ Chromabond™ Glass Fiber Frit are designed for individual packing of empty glass SPE columns with Chromabond™ adsorbents (glass columns, adsorbents sold separately).
Frits act as filters in solid phase extraction chromatography. They help to hold the column packing in place and facilitates sorbent packing in the column. Glass fiber columns/frits have many advantages:
  • Resistant to acids, bases and most organic solvents
  • Has low absorption for most biological macromolecules
  • Ideal for high-temperature applications
Specifications
For Use With (Application) Solid Phase Extraction
For Use With (Equipment) Glass Column
Type Frit
Volume (Metric) 6 mL
Material Glass Fiber
Quantity 250 Pack