Learn More
Macherey-Nagel™ Chromabond™ HR-P Polypropylene SPE Column
Product Code. 10536871 Shop All Macherey-Nagel Products
Click to view available options
Bed Weight:
100 mg
200 mg
500 mg
1 g
Column Capacity (Metric):
1 mL
3 mL
6 mL
Unit Size:
Each
Pack of 30
Product Code. 10536871
Brand: Macherey-Nagel™ 730111
Specifications
Column Capacity (Metric) 6 mL
Column Type SPE
Bed Weight 500 mg
Particle Size 50 to 100 μm
Type Solid Phase Extraction Column
Surface Area (Metric) 1200 m2/g