Learn More
Macherey-Nagel™ Chromabond™ HR-XA Polypropylene SPE Column
Brand: Macherey-Nagel™ 730954
Additional Details : Weight : 0.20000kg
Specifications
PS/DVB Copolymer | |
SPE | |
Solid Phase Extraction | |
Chromabond HR-XA | |
Spherical | |
850 m2/g |
3 mL | |
500 mg | |
85 μm | |
Solid Phase Extraction Column | |
55 to 65 Å |