Learn More
Macherey-Nagel™ CHROMABOND™ Complete Vacuum Manifold
Product Code. 10799172 Shop All Macherey-Nagel Products
Click to view available options
For Use With (Equipment):
up to 12 Columns
up to 16 LV Columns
up to 24 Columns
Unit Size:
Each
Product Code. 10799172
Brand: Macherey-Nagel™ 730150
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Designed for the simultaneous preparation of up to 12, 16 or 24 samples. Macherey-Nagel™ CHROMABOND™ Complete Vacuum Manifold handles columns and cartridges used in solid phase extraction (SPE).
The complete CHROMABOND Vacuum Manifold consists of:
  • Rectangular glass cabinet with polypropylene lid and lid gasket (2 sizes available):
    • Small holds up to 12 CHROMABOND™ columns or CHROMAFIX™ cartridges
    • Large holds up to 16 CHROMABOND™ LV columns or up to 24 CHROMABOND™ columns or CHROMAFIX™ cartridges
  • Removable needles on lower side of the lid
  • Vacuum gauge for reading the vacuum pressure in the chamber
  • Control valve for adjustment of the vacuum in the chamber
  • Replaceable valves with caps that allow individual vacuum control of each SPE column
  • Variable rack with exchangeable partitions, which accept a wide variety of vessels like test tubes, measuring flasks, scintillation vials, autosampler vials, plastic vials, etc.
Specifications
For Use With (Application) Solid Phase Extraction
For Use With (Equipment) up to 12 Columns
Type Vacuum Manifold
Quantity 1 Each