Learn More
Thermo Scientific™ Chromacol Vial Racks
Product Code. 10769304
Click to view available options
Holds:
Vials
Vials (100)
Vials (104)
Vials (105)
Material:
Aluminum
Foam
Plastic
For Use With (Equipment):
11 mm OD Vials
12 mm OD Vials
15 mm OD Vials
22 to 23mm OD Vials
22-23 mm OD Vials
8 mm OD Vials
Unit Size:
Each
Pack of 5
Product Code. 10769304
Brand: Thermo Scientific™ T10/20
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Ensure a safe working position on the lab bench and during transport with these robust vial racks.
Save space in the fridge while ensuring a safe working position on the bench and during transport. Available in a various materials, colors and capacities, Thermo Scientific™ Chromacol Vial Racks meet a variety of application needs
Choose from an assortment of chemically resistant and fairly robust racks.
  • Aluminum and plastic models are temperature resistant from -90 to +121°C (-130 to +250°F)
  • Aluminum and plastic models are autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vial Rack
Description Vial Rack
Autoclavable Autoclavable
Color White
Holds Vials
Material Plastic
Positions 10
For Use With (Equipment) 22 to 23mm OD Vials
Limited warranty period:
Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.