missing translation for 'onlineSavingsMsg'
Learn More

chroman-8-carbonyl chloride, Thermo Scientific™

Product Code. p-4391270
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10653455

Brand: Thermo Scientific Maybridge CC76102CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1034566-09-0
Molecular Formula C10H9ClO2
Molecular Weight (g/mol) 196.63
InChI Key TTYLUAYKWAEQDM-UHFFFAOYSA-N
Synonym chroman-8-carbonyl chloride, 3,4-dihydro-2h-1-benzopyran-8-carbonyl chloride, chromane-8-carbonyl chloride
PubChem CID 15480465

Specifications

Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 196.63
Percent Purity 90%
Chemical Name or Material chroman-8-carbonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.