Learn More
Metro™ Chrome Steel Shelves
Click to view available options
Dimensions (L x W):
1220 x 610 mm
1825 x 610 mm
Unit Size:
Each
Specifications
Specifications
| Product Type | Shelf |
| Material | Chrome Steel |
| Diameter (Metric) | 610 mm |
| Depth (Metric) | 610 mm |
| Dimensions (L x W) | 1825 x 610 mm |
| For Use With (Application) | For storage in ventilated area, the air flow is not disturbed |
| Height (Metric) | 189.5 cm |
| Length (Metric) | 182.50 cm |
| Load Bearing Capacity (Metric) | 270 kg |
| Shelf Type | Complete Stationary Unit 5 |
| Show More |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur