missing translation for 'onlineSavingsMsg'
Learn More

Chromogranin A Ab-2 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Epredia™ Chromogranin A Ab-2, Mouse Monoclonal Antibody.

127.50€ - 391.68€

Specifications

Antigen Chromogranin A Ab-2
Clone PHE5
Applications Immunofluorescence
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 6
Product Code Brand Form Quantity Price Quantity & Availability  
Product Code Brand Form Quantity Price Quantity & Availability  
12661257
View Documents Promotion Details
Epredia
MS-381-P1
Concentrated 500 μL
262.00€
500µL
View alternative products
Save Up to 
»
Estimated Shipment: 04-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
12621267
missing translation for 'viewDocuments' Promotion Details
Epredia
MS-381-R7
Prediluted 7 mL
193.00€
7mL
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 04-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
12651257
missing translation for 'viewDocuments' Promotion Details
Epredia
MS-381-PABX
Concentrated 200 μL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
12691257
missing translation for 'viewDocuments' Promotion Details
Epredia
MS-381-P0
Concentrated 100 μL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
12681257
missing translation for 'viewDocuments' Promotion Details
Epredia
MS-381-P
Concentrated 1 mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
12671257
missing translation for 'viewDocuments' Promotion Details
Epredia
MS-381-P1ABX
Concentrated 100 μL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
Description

Description

Chromogranin A (a protein of 439-amino acid which is encoded on chromosome 14) is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, pheo-chromocytomas, paragangliomas, and other neuro-endocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms.

Host Species: Mouse

Clone: PHE5

Isotype: IgG1

Species Reactivity: Human and Monkey. Does not react with rat. Others not known.

Immunogen: Human pheochromocytoma

Molecular Weight: 68-75 kDa

Positive Control: Pancreas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Chromogranin A Ab-2
Immunofluorescence
Unconjugated
RUO
Human, Non-human Primate
IgG1
PHE5
Monoclonal
Mouse
Oncogenesis
Human pheochromocytoma
Primary
Videos
SDS
Documents

Documents

Certificates