Learn More
Proteintech ChromoTek Binding Control Magnetic Agarose
Description
- Control of unspecific binding of proteins, DNA, etc. to beads
- Pre-clearing of cell lysate
- 20 reactions / 500 μL slurry
- Stored in 20% EtOH
Specifications
Specifications
| Quantity | 20 Reactions |
| Sufficient For | 20 Reactions |