Learn More
Proteintech ChromoTek mNeonGreen-Trap Agarose Beads
Product Code. 17343393
Click to view available options
Sufficient For:
10 Reactions
100 Reactions
20 Reactions
200 Reactions
400 Reactions
Unit Size:
10 reactions
100 reactions
20 reactions
200 reactions
400 reactions
Product Code. 17343393
Brand: Proteintech NTA400
mNeonGreen-Trap Agarose is an affinity reagent for IP of mNeonGreen-fusion proteins.
  • Consists of a mNeonGreen Nanobody/ VHH coupled to agarose beads
Specifications
Content And Storage Upon receipt store at 4°C. Do not freeze.
For Use With (Application) IP, CoIP, ChIP, RIP
Target mNeonGreen, Branchiostoma lanceolatum
Label or Dye Agarose
Species Alpaca
Sufficient For 400 Reactions