Learn More
Thermo Scientific Chemicals Chromotrope 2B
Catalog Number 11397706
Click to view available options
Quantity:
25 g
100 g
548-80-1
C16H9N3Na2O10S2
513.36
MFCD00003951
MQHFURHHKNKIRN-GVLVODDDSA-L
Acid Red 176; C.I. 16575
44135472
[Na+].[Na+].OC1=CC(=CC2=C1C(=O)\C(=N/NC1=CC=C(C=C1)[N+]([O-])=O)C(=C2)S([O-])(=O)=O)S([O-])(=O)=O
548-80-1
C16H9N3Na2O10S2
513.36
MFCD00003951
MQHFURHHKNKIRN-GVLVODDDSA-L
Acid Red 176; C.I. 16575
44135472
[Na+].[Na+].OC1=CC(=CC2=C1C(=O)\C(=N/NC1=CC=C(C=C1)[N+]([O-])=O)C(=C2)S([O-])(=O)=O)S([O-])(=O)=O
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point >300°C
Beilstein 4109733
Merck Index 14,2240
IUPAC Name disodium;5-hydroxy-3-[(4-nitrophenyl)hydrazinylidene]-4-oxonaphthalene-2,7-disulfonate
Formula Weight 513.37
Percent Purity 97%
Quantity 25 g
Chemical Name or Material Chromotrope 2B