missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cinnamyl alcohol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1000 g
Description
Cinnamyl alcohol is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used in perfumery and as well as a deodorant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cinnamyl alcohol is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used in perfumery and as well as a deodorant.
Solubility
Soluble in water 1.8 g/L (20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Incompatible with strong oxidizing agents.
Cinnamyl alcohol is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used in perfumery and as well as a deodorant.
Solubility
Soluble in water 1.8 g/L (20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Incompatible with strong oxidizing agents.
Specifications
Melting Point | 32°C to 35°C |
Density | 1.044 |
Boiling Point | 257°C to 258°C |
Flash Point | 126°C (258°F) |
Odor | Pleasant |
Linear Formula | C6H5CH=CHCH2OH |
Quantity | 250 g |
Beilstein | 1903999 |
Merck Index | 14,2302 |
Solubility Information | Soluble in water 1.8g/L (20°C). |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'