missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-3-Nonen-1-ol, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Used as flavoring agent for fresh, tropical, melon notes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as flavoring agent for fresh, tropical, melon notes.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from strong oxidizing agents.
Used as flavoring agent for fresh, tropical, melon notes.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from strong oxidizing agents.
Specifications
Density | 0.847 |
Boiling Point | 107°C to 109°C (20 mmHg) |
Flash Point | 95°C (203°F) |
Refractive Index | 1.45 |
Quantity | 5 g |
Solubility Information | Soluble in alcohol. Insoluble in water. |
Formula Weight | 142.24 |
Percent Purity | 97% |
Chemical Name or Material | cis-3-Nonen-1-ol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'