missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Hydroxy-L-proline, TRC

Product Code. 30389328
Change view
Click to view available options
Quantity:
50 mg
100 mg
Product Code. Quantity
30389328 50 mg
30401508 100 mg
2 options
This item is not returnable. View return policy

Product Code. 30389328

Brand: Toronto Research Chemicals TRCH95237550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 618-27-9
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.13
Synonym (4S)-4-Hydroxy-L-proline, (2S,4S)-4-Hydroxyproline, (S)-Allo-hydroxyproline, (2S,4S)-4-Hydroxypyrrolidine-2-carboxylic Acid, L-Allohydroxyproline, cis-4-Hydroxyproline, cis-Hydroxyproline
IUPAC Name (2S,4S)-4-hydroxypyrrolidine-2-carboxylic acid
SMILES O[C@@H]1CN[C@H](C(O)=O)C1

Specifications

InChI Formula InChI=1S/C5H9NO3/c7-3-1-4(5(8)9)6-2-3/h3-4,6-7H,1-2H2,(H,8,9)/t3-,4-/m0/s1
Quantity 50 mg
Formula Weight 131.0582
Shipping Condition Room Temperature
Chemical Name or Material cis-4-Hydroxy-L-proline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.